Skip to Content
Merck
All Photos(1)

Key Documents

739553

Sigma-Aldrich

Poly(styrene-block-methyl methacrylate)

average Mn 32,000, block ratio of 2:1

Synonym(s):

PS-PMMA, block copolymer, Poly(b-methyl methacrylate-polystyrene), PS-block-PMMA, PS-PMMA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002

form

solid

mol wt

average Mn 10,000 (PMMA)
average Mn 22,000 (polystyrene)
average Mn 32,000

transition temp

Tm 351-356 °C

Mw/Mn

<1.2

InChI

1S/C8H8.C5H8O2/c1-2-8-6-4-3-5-7-8;1-4(2)5(6)7-3/h2-7H,1H2;1H2,2-3H3

InChI key

ADFPJHOAARPYLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

22 kDa PS; 10 kDa PMMA; m ~200; n ~100

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel fabrication of 2D and 3D inverted opals and their application.
Jae Ah Lee et al.
Small (Weinheim an der Bergstrasse, Germany), 7(18), 2581-2586 (2011-08-03)
M H Geiger et al.
Clinical orthopaedics and related research, (382)(382), 258-266 (2001-01-12)
This controlled study compared the strength and porosity of 48 polymethylmethacrylate cement-implant constructs prepared with open bowl versus vacuum mix technique. Forty-eight blast finished stainless steel rods of 13 mm diameter were implanted with centralizers into 17-mm inner diameter tubes
S J Breusch et al.
Acta orthopaedica Scandinavica, 72(1), 29-35 (2001-05-01)
In a cadaver study, we prepared 29 paired human cadaver femora using 3 different broaches of identical geometry but different surface characteristics. In one group of 20 pairs, preparation with chipped-toothed broaches was compared to diamond-shaped broaches; in the other
F N K Kwong et al.
The Journal of bone and joint surgery. British volume, 88(1), 120-122 (2005-12-21)
The outcome of a cemented hip arthroplasty is partly dependent on the type of cement which is used. The production of an interface gap between the stem and the cement mantle as a result of shrinkage of the cement, may
Takeshi Ueki et al.
The journal of physical chemistry. B, 111(18), 4750-4754 (2007-02-27)
The solubility and phase behavior of poly(benzyl methacrylate) (PBzMA) and poly(styrene-co-methyl methacrylate) (P(St-co-MMA)) in a hydrophobic ionic liquid, 1-ethyl-3-methylimidazolium bis(trifluoromethane sulfone)imide ([C(2)mim][NTf(2)]), have been explored as a function of temperature. Although both polymers have solvatophobic phenyl groups and solvatophilic methacrylate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service