Skip to Content
Merck
All Photos(3)

Key Documents

738018

Sigma-Aldrich

2,7-Dibromo-9,9′-spirobifluorene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H14Br2
CAS Number:
Molecular Weight:
474.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

334-336 °C

SMILES string

Brc1ccc2-c3ccc(Br)cc3C4(c5ccccc5-c6ccccc46)c2c1

InChI

1S/C25H14Br2/c26-15-9-11-19-20-12-10-16(27)14-24(20)25(23(19)13-15)21-7-3-1-5-17(21)18-6-2-4-8-22(18)25/h1-14H

InChI key

UPJLZKCEPFAKSH-UHFFFAOYSA-N

General description

2,7-Dibromo-9,9′-spirobifluorene is a 9,9 substituted poly(2,7-fluorene) that is synthesized from 2,7-dribromo-9-fluorenone by reacting with a Grignard reagent of 2-bromobiphenyl. It has high photoluminescence and electroluminescent quantum efficiency that make it useful in the development of organic electronic devices.[1]

Application

2,7-Dibromo-9,9′-spirobifluorene can be used in the preparation of 9,9′-spirobifluorene-based small molecules, which can further be utilized as host materials in organic light emitting diodes (OLEDs).[2][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tri-, tetra-and pentamers of 9, 9?-spirobifluorenes through full ortho-linkage: High triplet-energy pure hydrocarbon host for blue phosphorescent emitter
Fan C, et al.
Organic Letters, 12(24), 5648-5651 (2010)
Syntheses and spectroscopic studies of spirobifluorene-bridged bipolar systems; photoinduced electron transfer reactions
Chien Y, et al.
Chemical Communications (Cambridge, England), 2874-2875 (2002)
Spiro-Functionalized Polyfluorene Derivatives as Blue Light-Emitting Materials
Yu W-L, et al.
Advanced Materials, 12(11), 828-831 (2000)
Shugo Sakaguchi et al.
The journal of physical chemistry. B, 122(36), 8614-8623 (2018-08-23)
Nanostructures composed of conjugated polymers or π-conjugated molecules provide sensing platforms with large specific surface areas. One of the feasible approaches to accessing such nanostructured miniaturized sensors with ultrahigh sensitivity is to develop a network of organic nanowires with optical/electronic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service