Skip to Content
Merck
All Photos(2)

Key Documents

737763

Sigma-Aldrich

Fmoc-Pro-Bt

0.95

Synonym(s):

(S)-(9H-Fluoren-9-yl)methyl 2-(1H-benzo[d][1,2,3]triazole-1-carbonyl)pyrrolidine-1-carboxylate, (S)-N-Fmoc-2-(benzotriazolylcarbonyl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22N4O3
CAS Number:
Molecular Weight:
438.48
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Fmoc-Pro-Bt, 95%

Assay

95%

form

solid

mp

163-168 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

O=C(OCC1c2ccccc2-c3ccccc13)N4CCC[C@H]4C(=O)n5nnc6ccccc56

InChI

1S/C26H22N4O3/c31-25(30-23-13-6-5-12-22(23)27-28-30)24-14-7-15-29(24)26(32)33-16-21-19-10-3-1-8-17(19)18-9-2-4-11-20(18)21/h1-6,8-13,21,24H,7,14-16H2/t24-/m0/s1

InChI key

ZTDUGEKWIJHWKX-DEOSSOPVSA-N

Application

Benzotriazole amino acids, or aminoacylbenzotriazolides, are versatile reagents for synthesizing peptides as well as their mimetics and conjugates. Benzotriazole amino acids have been used for the preparation of diverse derivatives including:

  • Polypeptidal benzotriazolides[1]
  • Peptidomimetics, such as aminoxypeptides[2], depsipeptides[3] and heterocyclic peptidomimetics[4]
  • Tagged peptides and peptidomimetics, particularly those with fluorescent labels[5][6]
  • N, O, S, and C linked peptide conjugates[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katritzky, A. R.; Angrish, P.; Todadze, E.
Synlett, 2392-2411 (2009)
Katritzky, A. R.; Narindoshvili, T.; Angrish, P.
Synthesis, 2013-2022 (2008)
Hansen, F. K.; Beagle, L. K.; Todadze, E.; Katritzky, A. R.
Heterocycles, 515-526 (2012)
Abdelmajeid, A.; Tala, S. R.; Amine, M. S.; Katritzky, A. R.
Synthesis, 2995-3005 (2011)
Alan R Katritzky et al.
The Journal of organic chemistry, 74(22), 8690-8694 (2009-10-17)
N-(Pg-alpha-aminoxy acids) 1a-g are converted to N-(Pg-alpha-aminoxyacyl)benzotriazoles 2a-g, which react under mild conditions with amines, alpha-amino acids/alpha-dipeptides, and alpha-aminoxy acids to give aminoxyacyl amides 3a-g, (3e+3e'), and (3g+3g'), aminoxy hybrid peptides 4a-h, (4a+4a'), 6a-d, 9a-e, (9a+9a'), and (9b+9b'), and alpha-aminoxy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service