Skip to Content
Merck
All Photos(1)

Key Documents

737321

Sigma-Aldrich

Triphenylbismuth

98% (99.9% trace metals basis)

Synonym(s):

TPB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi(C6H5)3
CAS Number:
Molecular Weight:
440.29
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98% (99.9% trace metals basis)

form

powder or crystals

reaction suitability

core: bismuth

availability

available only in USA

mp

76-81 °C

SMILES string

c1ccc(cc1)[Bi](c2ccccc2)c3ccccc3

InChI

1S/3C6H5.Bi/c3*1-2-4-6-5-3-1;/h3*1-5H;

InChI key

ZHXAZZQXWJJBHA-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Anjaneyulu et al.
Dalton transactions (Cambridge, England : 2003), 41(3), 1004-1012 (2011-11-23)
The reaction of triphenylbismuth [BiPh(3)] with several heterocyclic carboxylic acids was explored. Seven crystalline compounds, [PhBi(2-O(2)C-3-(OH)C(5)H(3)N)(2)(2-O(2)C-3-(OH)C(5)H(3)NH)] (5), [(Bi(2-O(2)C-3-(OH)C(5)H(3)N)(4))(C(5)H(5)NH)(C(5)H(5)N)] (7), [PhBi(2-O(2)C-C(4)H(3)N(2))(2)(2-O(2)C-C(4)H(4)N(2))·H(2)O] (8), [PhBi(2-O(2)C-C(9)H(6)N)(2)·H(2)O] (9), [Ph(2)Bi(O(2)C-C(4)H(3)O)] (10), [Ph(2)Bi(O(2)C-C(4)H(3)S)] (11) and [PhBi(O(2)C-C(4)H(3)S)(2)](2) (12), were prepared by simple reactions using BiPh(3) and the corresponding
Brad H Jones et al.
Macromolecular rapid communications, 42(4), e2000571-e2000571 (2020-12-11)
This communication describes a novel series of linear and crosslinked polyurethanes (PUs) and their selective depolymerization under mild conditions. Two unique polyols are synthesized bearing unsaturated units in a configuration designed to favor ring-closing metathesis (RCM) to five- and six-membered
Hu Xu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 75(1), 64-73 (2005-07-15)
Radiopaque cement containing barium sulfate causes significantly more bone resorption in vivo and in vitro than radiolucent cement. The aim of this study was to investigate the osteolytic potential of an alternative radiopaque agent, triphenyl bismuth (TPB). Bone cement particles
S N Abdulghani et al.
Journal of biomaterials science. Polymer edition, 14(11), 1229-1242 (2004-02-11)
Self-curing acrylic bone cements are widely used in the fixation of prosthetic implants in orthopaedic surgery. Commercial bone cements are rendered radiopaque by the addition of heavy metal salts of barium and zirconia. The addition of barium sulphate adversely affects
P A Mattie et al.
Journal of prosthodontics : official journal of the American College of Prosthodontists, 3(4), 213-218 (1994-12-01)
Current prosthetic acrylic resins are radiolucent and cannot be imaged using standard radiographic techniques. If accidentally impacted or ingested, delays in localizing or removing the foreign body may be life-threatening. The purpose of this study was to evaluate the influence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service