Skip to Content
Merck
All Photos(1)

Documents

735043

Sigma-Aldrich

Bis(chlorogold(I)) [1,1′-bis(diphenylphosphino)ferrocene]

95%

Synonym(s):

[μ-Ferrocene-1,1′-diylbis(diphenylphosphine-κP)]bis[chlorogold(I)]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H28Au2Cl2FeP2
CAS Number:
Molecular Weight:
1019.22
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

95%

form

solid

reaction suitability

core: gold
reagent type: catalyst
core: gold

mp

223-229 °C

SMILES string

[Fe].Cl[Au].Cl[Au].[CH]1[CH][CH][C]([CH]1)P(c2ccccc2)c3ccccc3.[CH]4[CH][CH][C]([CH]4)P(c5ccccc5)c6ccccc6

InChI

1S/2C17H14P.2Au.2ClH.Fe/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;;;;;/h2*1-14H;;;2*1H;/q;;2*+1;;;/p-2

InChI key

XJPNFMAYIDODRO-UHFFFAOYSA-L

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service