Skip to Content
Merck
All Photos(1)

Documents

733962

Sigma-Aldrich

2-(Difluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F2N
CAS Number:
Molecular Weight:
129.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

solubility

1.454 at 20 °C

density

1.204 g/mL at 25 °C

SMILES string

FC(F)c1ccccn1

InChI

1S/C6H5F2N/c7-6(8)5-3-1-2-4-9-5/h1-4,6H

InChI key

KUHSAAHTEMAJTF-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.1 °F

Flash Point(C)

41.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service