Skip to Content
Merck
All Photos(1)

Key Documents

733431

Sigma-Aldrich

3-Methylphenethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

density

0.939 g/mL at 25 °C

SMILES string

Cc1cccc(CCN)c1

InChI

1S/C9H13N/c1-8-3-2-4-9(7-8)5-6-10/h2-4,7H,5-6,10H2,1H3

InChI key

GUERDLPJJJMIEU-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service