Skip to Content
Merck
All Photos(2)

Key Documents

730459

Sigma-Aldrich

(2S,2′S-(−)-[N,N′-Bis(2-pyridylmethyl)]-2,2′-bipyrrolidinebis(acetonitrile)iron(II) hexafluoroantimonate

Synonym(s):

Fe(S,S-PDP), Iron(II) (S,S-(2-({(S)-2-[(S)-1-(pyridin-2-ylmethyl)pyrrolidin-2-yl]pyrrolidin-1-yl}methyl)pyridine)(bis-acetonitrile) hexafluoroantimonate, White Catalyst, [Fe(S,S-PDP)(CH3CN)2](SbF6)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32F12FeN6Sb2
CAS Number:
Molecular Weight:
931.90
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

optical activity

[α]20/D −30.0 to −20.0°, c = 0.2 in acetonitrile

reaction suitability

core: iron
reagent type: catalyst
reaction type: C-H Activation

mp

>300 °C

SMILES string

[Fe++].CC#N.CC#N.F[Sb-](F)(F)(F)(F)F.F[Sb-](F)(F)(F)(F)F.C1C[C@H]([C@H]2CCCN2Cc3ccccn3)N(C1)Cc4ccccn4

InChI

1S/C20H26N4.2C2H3N.12FH.Fe.2Sb/c1-3-11-21-17(7-1)15-23-13-5-9-19(23)20-10-6-14-24(20)16-18-8-2-4-12-22-18;2*1-2-3;;;;;;;;;;;;;;;/h1-4,7-8,11-12,19-20H,5-6,9-10,13-16H2;2*1H3;12*1H;;;/q;;;;;;;;;;;;;;;+2;2*+5/p-12/t19-,20-;;;;;;;;;;;;;;;;;/m1................./s1

InChI key

XUNWHBGEOCZDBX-QSKJABRXSA-B

Application

Catalyst for greener ketone synthesis by selective C-H oxidation of methylenes.

Combined Effects on Selectivity in Fe-Catalyzed Methylene Oxidation
Catalyst for:
  • Oxidative radical reactions
  • Selective oxidation of alkyl carbon-hydrogen bonds
  • Methylene group oxidation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service