Skip to Content
Merck
All Photos(2)

Key Documents

728233

Sigma-Aldrich

4-Chloro-2-butyn-1-ol

≥95.0% (GC)

Synonym(s):

1-Chloro-4-hydroxy-2-butyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5ClO
CAS Number:
Molecular Weight:
104.53
Beilstein:
1098382
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

storage temp.

−20°C

SMILES string

OCC#CCCl

InChI

1S/C4H5ClO/c5-3-1-2-4-6/h6H,3-4H2

InChI key

RPUJAOFSZSEVPC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service