Skip to Content
Merck
All Photos(1)

Key Documents

727083

Sigma-Aldrich

(S)-(+)-1-Aminoindan

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-(+)-1-Indanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
Beilstein:
2206708
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.562 (lit.)

bp

96-97 °C/8 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

N[C@H]1CCc2ccccc12

InChI

1S/C9H11N/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4,9H,5-6,10H2/t9-/m0/s1

InChI key

XJEVHMGJSYVQBQ-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service