Skip to Content
Merck
All Photos(1)

Key Documents

726907

Sigma-Aldrich

(R)-(−)-2-Amino-3-methylbutane

ChiPros®, produced by BASF, 99%

Synonym(s):

(R)-(−)-3-Methyl-2-butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13N
CAS Number:
Molecular Weight:
87.16
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

produced by BASF

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

density

0.75 g/mL at 20 °C (lit.)

SMILES string

CC(C)[C@@H](C)N

InChI

1S/C5H13N/c1-4(2)5(3)6/h4-5H,6H2,1-3H3/t5-/m1/s1

InChI key

JOZZAIIGWFLONA-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<69.8 °F

Flash Point(C)

< 21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service