Skip to Content
Merck
All Photos(1)

Key Documents

725285

Sigma-Aldrich

(Iodomethyl)triphenylphosphonium iodide

95%

Synonym(s):

NSC 116665

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17I2P
CAS Number:
Molecular Weight:
530.12
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

260-266 °C

functional group

iodo
phosphine

SMILES string

[I-].IC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H17IP.HI/c20-16-21(17-10-4-1-5-11-17,18-12-6-2-7-13-18)19-14-8-3-9-15-19;/h1-15H,16H2;1H/q+1;/p-1

InChI key

NAVMMSRRNOXQMJ-UHFFFAOYSA-M

Application

Reactant or reagent for synthesis of:
  • Phosphacyclic compounds via oxidative dimerizations
  • Enamides via cross-coupling
  • Cyclopeptide alkaloids for use as antibacterial or cytotoxic agents
  • Terminal alkynes via dehydrohalogenation of aldehydes

Reactant for vinylcyclopropanation / cyclopentenation of strained alkenes using a sequential carborhodation process

Used as an olefination agent in the Lewis acid catalysis of electrocyclization of trienes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service