Skip to Content
Merck
All Photos(2)

Key Documents

722049

Sigma-Aldrich

7-Bromo-8-hydroxyquinoline

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$319.00

R$319.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$319.00

About This Item

Empirical Formula (Hill Notation):
C9H6BrNO
CAS Number:
Molecular Weight:
224.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$319.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

138-143 °C

functional group

bromo

SMILES string

Oc1c(Br)ccc2cccnc12

InChI

1S/C9H6BrNO/c10-7-4-3-6-2-1-5-11-8(6)9(7)12/h1-5,12H

InChI key

ICKPMTNGWVNOGC-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rungrot Cherdtrakulkiat et al.
EXCLI journal, 18, 445-453 (2019-08-20)
Antimicrobial resistance has become a prime global concern. An ability of the microbes to produce enzymes to destroy antimicrobial drugs is one of the well-known mechanisms underlying the resistance. 8-Hydroxyquinoline (8HQ) and derivatives were reported to exert diverse biological effects
Dmytro Havrylyuk et al.
European journal of medicinal chemistry, 156, 790-799 (2018-07-29)
8-Hydroxyquinolines (HQ), including clioquinol, possess cytotoxic properties and are widely used as ligands for metal-based anticancer drug research. The number and identity of substituents on the HQ can have a profound effect on activity for a variety of inorganic compounds.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service