Skip to Content
Merck
All Photos(1)

Key Documents

721980

Supelco

Ethylene glycol solution

NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07
MDL number:
UNSPSC Code:
12352137
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

form

solution

concentration

80% in DMSO-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

High temperature calibration

Quantity

3 mm O.D. tube contains 0.235 mL.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Peter Mygind Leth et al.
Forensic science international, 155(2-3), 179-184 (2005-10-18)
Ethylene glycol (EG) can be found in many agents, such as antifreeze. Ingestion of EG may cause serious poisoning. Adults are typically exposed when EG is ingested as a cheap substitute for ethanol or in suicide-attempts. Children may be exposed
Andrea Marchionni et al.
ChemSusChem, 6(3), 518-528 (2013-02-14)
The electrooxidation of ethylene glycol (EG) and glycerol (G) has been studied: in alkaline media, in passive as well as active direct ethylene glycol fuel cells (DEGFCs), and in direct glycerol fuel cells (DGFCs) containing Pd-(Ni-Zn)/C as an anode electrocatalyst
Mona Sheikhi et al.
Fertility and sterility, 100(1), 170-177 (2013-04-10)
To study the preservation of follicles within ovarian tissue vitrified using only one or a combination of three permeating cryoprotectants. Experimental study. University hospital. Ovarian tissue was donated by consenting women undergoing elective cesarean section. Ovarian tissue was vitrified in
Yi Wang et al.
Journal of the American Chemical Society, 135(5), 1941-1951 (2013-01-16)
This article describes a robust method for the facile synthesis of small Ag nanocubes with edge lengths controlled in the range of 18-32 nm. The success of this new method relies on the substitution of ethylene glycol (EG)--the solvent most
Revital Goldshmid et al.
Methods (San Diego, Calif.), 84, 35-43 (2015-05-02)
A method for the preparation of suspension culture microcapsules used in the bioprocessing of human mesenchymal stem cells (hMSCs) is reported. The microcapsules are prepared from a semi-synthetic hydrogel comprising Pluronic®F127 conjugated to denatured fibrinogen. The Pluronic-fibrinogen adducts display a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service