Skip to Content
Merck
All Photos(1)

Key Documents

721727

Sigma-Aldrich

1-Boc-3-bromopiperidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,063.00

R$1,063.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$1,063.00

About This Item

Empirical Formula (Hill Notation):
C10H18BrNO2
CAS Number:
Molecular Weight:
264.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,063.00


Check Cart for Availability

Request a Bulk Order

form

liquid

refractive index

n/D 1.488

density

1.270 g/L at 25 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

BrC1CN(CCC1)C(=O)OC(C)(C)C

InChI

1S/C10H18BrNO2/c1-10(2,3)14-9(13)12-6-4-5-8(11)7-12/h8H,4-7H2,1-3H3

InChI key

YCUDHDNCZHPAJK-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service