Skip to Content
Merck
All Photos(2)

Key Documents

721085

Sigma-Aldrich

Zinc oxide, dispersion

nanoparticles, 40 wt. % in ethanol, <130 nm particle size

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles

concentration

40 wt. % in ethanol

particle size

<130 nm

density

1.25 g/mL±0.15 g/mL at 25 °C

SMILES string

[Zn]=O

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.0 °F

Flash Point(C)

12.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

The influence of 1D, meso-and crystal structures on charge transport and recombination in solid-state dye-sensitized solar cells
Docampo P, et al.
Journal of Material Chemistry A, 1(39), 12088-12095 (2013)
Ting Cheng et al.
Acta biomaterialia, 54, 186-200 (2017-03-21)
Near-infrared (NIR) triggered chemical delivery allows on-demand release with the advantage of external tissue stimulation. Bioresorbable polyester poly-l-lactic acid (PLLA) was compounded with photoadditives of neat zinc oxide (ZnO) nanoparticles and 980→365nm LiYF Upconverting nanoparticles and zinc oxide nanorods were
Characterizing the Structure and Defect Concentration of ZnO Nanoparticles in a Colloidal Solution.
Rossi T, et al.
The Journal of Physical Chemistry C, 118(33), 19422-19430 (2014)
Fadri Gottschalk et al.
Environmental pollution (Barking, Essex : 1987), 181, 287-300 (2013-07-17)
Scientific consensus predicts that the worldwide use of engineered nanomaterials (ENM) leads to their release into the environment. We reviewed the available literature concerning environmental concentrations of six ENMs (TiO2, ZnO, Ag, fullerenes, CNT and CeO2) in surface waters, wastewater
Rizwan Wahab et al.
Journal of biomedical nanotechnology, 9(7), 1181-1189 (2013-08-06)
This paper reports the synthesis and characterization of ZnO nanoparticles prepared by soft chemical process. The nanoparticles of ZnO possess wurtzite hexagonal phase and were used for the induction of cell death in malignant human T98G gliomas, KB epithermoids and

Articles

Operation principle and market dominance of single crystalline silicon solar cells.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service