Skip to Content
Merck
All Photos(3)

Key Documents

718181

Sigma-Aldrich

N-4-Methanesulfonamidephenylboronic acid

≥95%

Synonym(s):

4-(Dihydroxyboryl)-N-(methylsulfonyl)phenylamine, 4-(Methanesulfonylamino)phenylboronic acid, 4-Methylsulfonylaminophenylboronic acid, [4-[(Methanesulfonyl)amino]phenyl]boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,147.00

R$1,147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$1,147.00

About This Item

Empirical Formula (Hill Notation):
C7H10BNO4S
CAS Number:
Molecular Weight:
215.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

R$1,147.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

solid

SMILES string

CS(=O)(=O)Nc1ccc(cc1)B(O)O

InChI

1S/C7H10BNO4S/c1-14(12,13)9-7-4-2-6(3-5-7)8(10)11/h2-5,9-11H,1H3

InChI key

NDVJJEADFLTFCD-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Potent and selective inhibitors of IKK-β
  • Inhibitors for treatment of osteoporosis
  • Inhibitors of human farnesyl pyrophosphate synthase
  • Suzuki-type Pd(0) coupling reactions in the synthesis of 2-arylpurines as Cdk inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service