715107
Methyl 2-aminopyridine-3-carboxylate
97%
Synonym(s):
Methyl 2-aminonicotinate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
mp
82-86 °C
SMILES string
COC(=O)c1cccnc1N
InChI
1S/C7H8N2O2/c1-11-7(10)5-3-2-4-9-6(5)8/h2-4H,1H3,(H2,8,9)
InChI key
NZZDEODTCXHCRS-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dalton transactions (Cambridge, England : 2003), 49(14), 4315-4322 (2020-03-13)
This study tries to provide new solutions to increase the efficiency of conversion of photons in solar cells, using photoluminescent Cu(i) coordination polymers (CPs) as possible alternative materials of lower cost, than those used today, based on lanthanides. The selected
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service