Skip to Content
Merck
All Photos(1)

Documents

714852

Sigma-Aldrich

1,8-Diazabicyclo[5.4.0]undec-7-ene solution

1 M in THF

Synonym(s):

DBU solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N2
Molecular Weight:
152.24
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

concentration

1 M in THF

refractive index

n20/D 1.425

density

0.899 g/mL at 25 °C

SMILES string

C1CCN2CCCN=C2CC1

InChI

1S/C9H16N2/c1-2-5-9-10-6-4-8-11(9)7-3-1/h1-8H2

InChI key

GQHTUMJGOHRCHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.0 °F

Flash Point(C)

-17.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Samorì et al.
Bioresource technology, 101(9), 3274-3279 (2010-01-15)
Lipid extraction is a critical step in the development of biofuels from microalgae. Here a new procedure was proposed to extract hydrocarbons from dried and water-suspended samples of the microalga Botryococcus braunii by using switchable-polarity solvents (SPS) based on 1,8-diazabicyclo-[5.4.0]-undec-7-ene
Wanli Chen et al.
The Journal of organic chemistry, 77(3), 1585-1591 (2012-01-19)
DBU-promoted cyclization of ortho-(3-hydroxy-1-alkynyl)benzamide is presented, providing an efficient method for the synthesis of trans-3,4-dihydroisoquinolin-1(2H)-ones and (E)-4-(1-alkenyl)isoquinolin-1(2H)-ones under mild conditions.
Highly efficient organosuperbase-catalyzed Mannich-type reactions of sulfonylimidates with imines: successful use of aliphatic imines as substrates and a unique reaction mechanism.
Junya Nakano et al.
Angewandte Chemie (International ed. in English), 51(38), 9525-9529 (2012-08-30)
Wim L Noorduin et al.
Angewandte Chemie (International ed. in English), 48(51), 9600-9606 (2009-12-01)
A century ago Wilhelm Ostwald received the Nobel Prize for Chemistry. Although Ostwald was never significantly involved with the phenomenon of chirality, one of his discoveries, Ostwald ripening, is thought to be involved in a recently discovered method in which
Lun-Zhi Dai et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(13), 3701-3706 (2007-01-20)
Reactions of salicylaldehydes with ethyl buta-2,3-dienoate or penta-3,4-dien-2-one catalyzed by a catalytic amount of potassium carbonate produce the corresponding 2 H-chromene products in moderate to good yields under mild conditions. A plausible reaction mechanism is discussed in the light of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service