Skip to Content
Merck
All Photos(1)

Key Documents

712620

Sigma-Aldrich

N-Boc-4-azido-L-homoalanine (dicyclohexylammonium) salt

≥97% (CE)

Synonym(s):

(S)-4-Azido-2-(Boc-amino)butyric acid (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H39N5O4
CAS Number:
Molecular Weight:
425.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (CE)

form

crystals

optical activity

[α]/D +15.5±1.0°, c = 1 in ethanol

composition

carbon, 57.5-61.0%
nitrogen, 16.0-16.9%

reaction suitability

reaction type: click chemistry

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)N[C@@H](CCN=[N+]=[N-])C(O)=O

InChI

1S/C12H23N.C9H16N4O4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-9(2,3)17-8(16)12-6(7(14)15)4-5-11-13-10/h11-13H,1-10H2;6H,4-5H2,1-3H3,(H,12,16)(H,14,15)/t;6-/m.0/s1

InChI key

UCLAYMPJMDBLOX-ZCMDIHMWSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A James Link et al.
Journal of the American Chemical Society, 126(34), 10598-10602 (2004-08-26)
An improved protocol for copper-catalyzed triazole formation on the bacterial cell surface is described. Addition of highly pure CuBr to cells treated with azidohomoalanine (2) leads to ca. 10-fold more extensive cell surface labeling than previously observed. This highly active

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service