Skip to Content
Merck
All Photos(1)

Key Documents

712558

Sigma-Aldrich

Methoxypolyethylene glycol maleimide

750

Synonym(s):

Polyethylene glycol, MeO-PEG-Mal, PEG-maleimide, mono-Methyl polyethylene glycol 2-maleimidoethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$1,905.00

R$1,905.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
R$1,905.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

R$1,905.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

mol wt

PEG average Mn 750

Ω-end

maleimide

α-end

methoxy

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Circulatory half-life is a key success factor for new drugs. In this respect, PEGylation or PEG-ing—the modification of potential candidates ranging from non-peptidic small molecules to peptides and proteins, antibody fragments, aptamers, and saccharides or oligonucleotides with polyethylene glycol chains—offers numerous advantages.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service