Skip to Content
Merck
All Photos(2)

Key Documents

712280

Sigma-Aldrich

(R)-Tetrahydro-3-furoic acid

≥97.0%

Synonym(s):

(R)-Tetrahydro-3-furancarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$5,072.00

R$5,072.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$5,072.00

About This Item

CAS Number:
Beilstein:
1281202
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

R$5,072.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (GC)
≥97.0%
97.0-103.0% (T)

form

liquid

optical purity

enantiomeric excess: ≥95.0%

functional group

carboxylic acid
ether

SMILES string

OC(=O)[C@@H]1CCOC1

InChI

1S/C5H8O3/c6-5(7)4-1-2-8-3-4/h4H,1-3H2,(H,6,7)/t4-/m1/s1

InChI key

BOTREHHXSQGWTR-SCSAIBSYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service