Skip to Content
Merck
All Photos(1)

Key Documents

710091

Sigma-Aldrich

3-(1,3-Dioxolan-2-yl)phenylboronic acid pinacol ester

Synonym(s):

2-(3-(1,3-dioxolan-2-yl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21BO4
CAS Number:
Molecular Weight:
276.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

45-50 °C

functional group

ether

SMILES string

CC1(C)OB(OC1(C)C)c2cccc(c2)C3OCCO3

InChI

1S/C15H21BO4/c1-14(2)15(3,4)20-16(19-14)12-7-5-6-11(10-12)13-17-8-9-18-13/h5-7,10,13H,8-9H2,1-4H3

InChI key

SVIZIDROXRZRQS-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service