708518
1,3-Bis(diphenylphosphinomethyl)benzene
Synonym(s):
α,α′-Bis(diphenylphosphino)-m-xylene, [1,3-Phenylenebis(methylene)]bis[diphenyl]phosphine
About This Item
Recommended Products
form
powder
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
50-55 °C
functional group
phosphine
SMILES string
C(P(c1ccccc1)c2ccccc2)c3cccc(CP(c4ccccc4)c5ccccc5)c3
InChI
1S/C32H28P2/c1-5-16-29(17-6-1)33(30-18-7-2-8-19-30)25-27-14-13-15-28(24-27)26-34(31-20-9-3-10-21-31)32-22-11-4-12-23-32/h1-24H,25-26H2
InChI key
QLSOZHFOFHARBJ-UHFFFAOYSA-N
Application
- Cyclometalation of pincer ligands
- Insertion of carbon dioxide into (PCP)PDII-Me Bonds
- Synthesis of iridium complexes containing bidentate phosphine ligands
- Transcyclometalation processes
- Synthesis of vinylidene and carbyne complexes
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service