Skip to Content
Merck
All Photos(1)

Key Documents

70750

Sigma-Aldrich

Dichloro(chloromethyl)methylsilane

Wacker Chemie AG, ≥98.0% (GC)

Synonym(s):

Silane CMM1, (Chloromethyl)dichloromethylsilane, (Chloromethyl)methyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2)Si(CH3)Cl2
CAS Number:
Molecular Weight:
163.51
Beilstein:
969205
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

≥98.0% (GC)

form

liquid

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.449 (lit.)

bp

121-122 °C (lit.)

density

1.284 g/mL at 25 °C (lit.)

SMILES string

C[Si](Cl)(Cl)CCl

InChI

1S/C2H5Cl3Si/c1-6(4,5)2-3/h2H2,1H3

InChI key

JAYBZWYBCUJLNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugen Weisheim et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(35), 12436-12448 (2015-07-28)
Directed tridentate Lewis acids based on the 1,3,5-trisilacyclohexane skeleton with three ethynyl groups [CH2Si(Me)(C2H)]3 were synthesised and functionalised by hydroboration with HB(C6F5)2, yielding the ethenylborane {CH2Si(Me)[C2H2B(C6F5)2]}3, and by metalation with gallium and indium organyls affording {CH2Si(Me)[C2M(R)2]}3 (M = Ga, In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service