Skip to Content
Merck
All Photos(1)

Key Documents

705934

SAFC

N-Cbz-cis-4-Hydroxy-L-proline methyl ester

97%

Synonym(s):

Methyl (2S,4S)-N-Cbz-4-hydroxypyrrolidine-2-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$919.00

R$919.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 G
R$919.00

About This Item

Empirical Formula (Hill Notation):
C14H17NO5
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

R$919.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Assay

97%

form

solid

mp

80-85 °C

SMILES string

COC(=O)[C@@H]1C[C@H](O)CN1C(=O)OCc2ccccc2

InChI

1S/C14H17NO5/c1-19-13(17)12-7-11(16)8-15(12)14(18)20-9-10-5-3-2-4-6-10/h2-6,11-12,16H,7-9H2,1H3/t11-,12-/m0/s1

InChI key

VVKAGQHUUDRPOI-RYUDHWBXSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service