Skip to Content
Merck
All Photos(1)

Key Documents

705225

Sigma-Aldrich

7-Methylxanthine-2,4,5,6-13C4, 1,3-15N2 (with variable 15N labeling at N9)

≥98 atom %, ≥98% (CP)

Synonym(s):

2,6-Dihydroxy-7-methylpurine-2,4,5,6-13C4, 1,3-15N2 (partial 15N labeling at N9)

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
R$4,017.00
5 MG
R$10,280.00

R$4,017.00


Estimated to ship onMay 22, 2025



Select a Size

Change View
2 MG
R$4,017.00
5 MG
R$10,280.00

About This Item

Empirical Formula (Hill Notation):
13C4C2H615N2N2O2
Molecular Weight:
172.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

R$4,017.00


Estimated to ship onMay 22, 2025


isotopic purity

≥98 atom %

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

≥300 °C

mass shift

M+7

storage temp.

−20°C
−20°C

SMILES string

Cn1c[15n][13c]2[15NH][13C](=O)[15NH][13C](=O)[13c]12

InChI

1S/C6H6N4O2/c1-10-2-7-4-3(10)5(11)9-6(12)8-4/h2H,1H3,(H2,8,9,11,12)/i3+1,4+1,5+1,6+1,7+1,8+1,9+1

InChI key

PFWLFWPASULGAN-JVYICBGUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service