Skip to Content
Merck
All Photos(1)

Key Documents

704571

Sigma-Aldrich

Bis(8-hydroxy-2-methylquinoline)-(4-phenylphenoxy)aluminum

95%

Synonym(s):

BAlq

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H25AlN2O3
CAS Number:
Molecular Weight:
512.53
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

form

powder

mp

207-214 °C

λmax

259 nm

fluorescence

λem 334, 477 nm in THF

OLED Device Performance

ITO/CuPc/NPD/CBP:Firpic (6%)/BAlq3/LiF/Al

  • Color: blue
  • Max. EQE: 4.5 %

SMILES string

Cc1ccc2cccc(O[Al](Oc3ccc(cc3)-c4ccccc4)Oc5cccc6ccc(C)nc56)c2n1

InChI

1S/C12H10O.2C10H9NO.Al/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10;2*1-7-5-6-8-3-2-4-9(12)10(8)11-7;/h1-9,13H;2*2-6,12H,1H3;/q;;;+3/p-3

InChI key

UFVXQDWNSAGPHN-UHFFFAOYSA-K

Application

Electron Transport Layer (ETL) material

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent progress of molecular organic electroluminescent materials and devices
Hung, L.S.; et al.
Materials Science & Engineering. R, Reports : A Review Journal, 39, 143 - 222 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service