Skip to Content
Merck
All Photos(1)

Key Documents

704318

Sigma-Aldrich

4,4′-Biphenyldiboronic acid bis(neopentyl) ester

97%

Synonym(s):

4,4′-Bis-(5,5-dimethyl-[1,3,2]dioxaborinane-2-yl)-biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28B2O4
CAS Number:
Molecular Weight:
378.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

powder

mp

241-245 °C

SMILES string

CC1(C)COB(OC1)c2ccc(cc2)-c3ccc(cc3)B4OCC(C)(C)CO4

InChI

1S/C22H28B2O4/c1-21(2)13-25-23(26-14-21)19-9-5-17(6-10-19)18-7-11-20(12-8-18)24-27-15-22(3,4)16-28-24/h5-12H,13-16H2,1-4H3

InChI key

NJXUWONHNYJPMY-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service