Skip to Content
Merck
All Photos(1)

Documents

701629

Sigma-Aldrich

Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24F6RhSb
CAS Number:
Molecular Weight:
555.02
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

mp

58-60 °C

storage temp.

−20°C

SMILES string

[Rh+].F[Sb-](F)(F)(F)(F)F.C1CC=CCCC=C1.C2CC=CCCC=C2

InChI

1S/2C8H12.6FH.Rh.Sb/c2*1-2-4-6-8-7-5-3-1;;;;;;;;/h2*1-2,7-8H,3-6H2;6*1H;;/q;;;;;;;;+1;+5/p-6/b2*2-1-,8-7-;;;;;;;;

InChI key

BROZUPPJWNODCU-SXGOCSEPSA-H

Application

Catalyst for:
  • [3+2] Cycloaddition
  • Preparation of diastereomeric chiral phosphorus mon- and bidentate diamidophosphite ligands based on dianydromannitol and their use in asymmetric metallocomplex catalysis
  • C-C bond-forming hydrogenation

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service