Skip to Content
Merck
All Photos(3)

Key Documents

701440

Sigma-Aldrich

2-(4-Methoxybenzyloxy)-4-methylquinoline

97%

Synonym(s):

Dudley Reagent II, PMBO-L, PMBO-lepidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO2
CAS Number:
Molecular Weight:
279.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

powder

mp

73-78 °C

SMILES string

COc1ccc(COc2cc(C)c3ccccc3n2)cc1

InChI

1S/C18H17NO2/c1-13-11-18(19-17-6-4-3-5-16(13)17)21-12-14-7-9-15(20-2)10-8-14/h3-11H,12H2,1-2H3

InChI key

NZVDQMQUHTVZAH-UHFFFAOYSA-N

Application

Dudley Reagents

  • Reagent for the protection of alcohols under relatively neutral and acidic reaction conditions

Legal Information

Sold under license from FSURF, US Patent Application Serial Nos. 60/827,763, 60/862,121 and related patents apply.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dudley Reagents provide mild protection for alcohols and carboxylic acids in synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service