Skip to Content
Merck
All Photos(1)

Documents

699020

Sigma-Aldrich

Trimethylaluminum solution

2.0 M in chlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Al
CAS Number:
Molecular Weight:
72.09
Beilstein:
3587197
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

concentration

2.0 M in chlorobenzene

density

1.034 g/mL at 25 °C

SMILES string

C[Al](C)C

InChI

1S/3CH3.Al/h3*1H3;

InChI key

JLTRXTDYQLMHGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

<-22.0 °F

Flash Point(C)

< -30 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahtab Farzin et al.
Pathology, 47(4), 302-307 (2015-05-06)
BRCA1-associated protein 1 (BAP1) is a tumour suppressor gene frequently inactivated in mesothelioma, rarely also in association with germline mutation. BAP1 mutations have been associated with improved prognosis and distinct clinicopathological features. We sought to determine the clinicopathological significance of
J L L Robinson et al.
Oncogene, 33(50), 5666-5674 (2013-12-03)
Castration-resistant prostate cancer (CRPC) continues to pose a significant clinical challenge with new generation second-line hormonal therapies affording limited improvement in disease outcome. As the androgen receptor (AR) remains a critical driver in CRPC, understanding the determinants of its transcriptional
Shahram Khosravi et al.
The Journal of investigative dermatology, 135(5), 1358-1367 (2015-01-07)
Human cutaneous melanoma is a devastating skin cancer because of its invasive nature and high metastatic potential. We used tissue microarray to study the role of human eukaryotic translation initiation factor 4E (eIF4E) in melanoma progression in 448 melanocytic lesions
Alexander C Kozen et al.
ACS nano, 9(6), 5884-5892 (2015-05-15)
Lithium metal is considered to be the most promising anode for next-generation batteries due to its high energy density of 3840 mAh g(-1). However, the extreme reactivity of the Li surface can induce parasitic reactions with solvents, contamination, and shuttled
J Stebbing et al.
Oncogene, 34(16), 2103-2114 (2014-06-10)
Kinase suppressor of Ras-1 (KSR1) facilitates signal transduction in Ras-dependent cancers, including pancreatic and lung carcinomas but its role in breast cancer has not been well studied. Here, we demonstrate for the first time it functions as a tumor suppressor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service