694908
Lanthanum
foil, thickness 1 mm, 99.9% (rare earth, metals basis)
About This Item
Assay
99.9% (rare earth, metals basis)
form
foil
reaction suitability
reagent type: catalyst
core: lanthanum
resistivity
54 μΩ-cm
thickness
1 mm
bp
3464 °C (lit.)
mp
920 °C (lit.)
density
6.19 g/mL at 25 °C (lit.)
SMILES string
[La]
InChI
1S/La
InChI key
FZLIPJUXYLNCLC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Quantity
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Water-react 1
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service