Skip to Content
Merck
All Photos(3)

Key Documents

694517

Sigma-Aldrich

QuadraPure® PHE

400-1100 μm particle size

Synonym(s):

QuadraPure® phenolic functionalized polystyrene

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
R$11,299.00

R$11,299.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
R$11,299.00

About This Item

UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

R$11,299.00


Check Cart for Availability

Request a Bulk Order

product line

QuadraPure®

Quality Level

form

beads

reaction suitability

reaction type: solution phase peptide synthesis

particle size

400-1100 μm

Application

Application: Metal scavenger: Fe(ΙΙΙ), Pd(ΙΙ), Rh(ΙΙΙ)

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service