Skip to Content
Merck
All Photos(2)

Documents

693804

Sigma-Aldrich

Zirconium-scandium-iron alloy

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr4Sc1Fe10
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative category

SMILES string

[Sc].[Fe].[Zr]

InChI

1S/Fe.Sc.Zr

InChI key

JYODYTROXJSIEE-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

High pressure vessels for mobile storage of hydrogen. New storage systems based on AB2-type intermetallics with good hydrogen absorption parameters. Materials are typical representative of AB2 alloys.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service