Skip to Content
Merck
All Photos(1)

Documents

692425

Sigma-Aldrich

(R)-RuCl[(p-cymene)(DTBM-SEGPHOS®)]Cl

Synonym(s):

Chloro[(R)−(−)5,5′−bis[bis(3,5−di−tert−butyl−4−methoxyphenyl)phosphino]−4,4′−bi−1,3−benzodioxole](p−cymene)ruthenium(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C84H114ClO8P2Ru]+Cl−
CAS Number:
Molecular Weight:
1485.72
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

powder

storage temp.

2-8°C

SMILES string

Cl[Ru]Cl.CC(C)c1ccc(C)cc1.COc2c(cc(cc2C(C)(C)C)P(c3cc(c(OC)c(c3)C(C)(C)C)C(C)(C)C)c4ccc5OCOc5c4-c6c7OCOc7ccc6P(c8cc(c(OC)c(c8)C(C)(C)C)C(C)(C)C)c9cc(c(OC)c(c9)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C74H100O8P2.C10H14.2ClH.Ru/c1-67(2,3)47-33-43(34-48(61(47)75-25)68(4,5)6)83(44-35-49(69(7,8)9)62(76-26)50(36-44)70(10,11)12)57-31-29-55-65(81-41-79-55)59(57)60-58(32-30-56-66(60)82-42-80-56)84(45-37-51(71(13,14)15)63(77-27)52(38-45)72(16,17)18)46-39-53(73(19,20)21)64(78-28)54(40-46)74(22,23)24;1-8(2)10-6-4-9(3)5-7-10;;;/h29-40H,41-42H2,1-28H3;4-8H,1-3H3;2*1H;/q;;;;+2/p-2

InChI key

MYYXUILRYSITRR-UHFFFAOYSA-L

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service