Skip to Content
Merck
All Photos(1)

Key Documents

691054

Sigma-Aldrich

1,3-Dihydroxyimidazolium bis(trifluoromethylsulfonyl)imide

98%

Synonym(s):

(OH)2Im-NTf2

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
R$8,111.00

R$8,111.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Select a Size

Change View
50 G
R$8,111.00

About This Item

Empirical Formula (Hill Notation):
C5H5F6N3O6S2
CAS Number:
Molecular Weight:
381.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$8,111.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (13C-NMR)
≥98% (19F-NMR)
≥98% (31P-NMR)
98%

form

liquid

refractive index

n20/D 1.4275

suitability

complies for IR spectroscopy

functional group

fluoro

SMILES string

On1cc[n+](O)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C3H5N2O2.C2F6NO4S2/c6-4-1-2-5(7)3-4;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h1-3,6-7H;/q+1;-1

InChI key

SWVAEQIBLYMHDX-UHFFFAOYSA-N

Application

Ionic liquid and NHC precatalyst[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N,N-Di(alkoxy)imidazolium salts: new patent-free ionic liquids and NHC-precatalysts
Laus, G., et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 62, 295-308 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service