Skip to Content
Merck
All Photos(2)

Documents

689017

Sigma-Aldrich

N-[(2S)-2-Pyrrolidinylmethyl]-trifluoromethanesulfonamide

≥98.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11F3N2O2S
CAS Number:
Molecular Weight:
232.22
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (T)

form

powder

optical activity

[α]/D +13±1°, c = 1 in methanol

optical purity

ee: ≥98.5%

SMILES string

FC(F)(F)S(=O)(=O)NC[C@@H]1CCCN1

InChI

1S/C6H11F3N2O2S/c7-6(8,9)14(12,13)11-4-5-2-1-3-10-5/h5,10-11H,1-4H2/t5-/m0/s1

InChI key

RIWFUAUXWIEOTK-YFKPBYRVSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liansuo Zu et al.
Organic letters, 8(14), 3077-3079 (2006-06-30)
[reaction: see text] A recycle and reusable fluorous (S)-pyrrolidine sulfonamide organocatalyst has been developed for promoting highly enantio- and diastereoselective Michael addition reactions of ketones and aldehydes with nitroolefins in water. The organocatalyst is conveniently recovered from the reaction mixtures
Liansuo Zu et al.
Organic letters, 10(6), 1211-1214 (2008-02-15)
Fluorous (S) pyrrolidine sulfonamide serves as an efficient promoter for highly enantioselective aldol reactions of ketones and aldehydes with aromatic aldehydes on water. A notable feature of the organocatalyst is that it can be recovered from the reaction mixtures by
Direct, highly enantioselective pyrrolidine sulfonamide catalyzed Michael addition of aldehydes to nitrostyrenes.
Wei Wang et al.
Angewandte Chemie (International ed. in English), 44(9), 1369-1371 (2005-01-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service