Skip to Content
Merck
All Photos(3)

Key Documents

686050

Sigma-Aldrich

Lithium amide

greener alternative

hydrogen-storage grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNH2
CAS Number:
Molecular Weight:
22.96
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

hydrogen-storage grade

Assay

95%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

380-400 °C

density

1.178 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li].CN

InChI

1S/Li.H2N/h;1H2/q+1;-1

InChI key

AFRJJFRNGGLMDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium amide (LiNH2) is an amide of an alkali metal with lithium atoms having larger positive charges than hydrogen atoms. It is a moisture-sensitive molecule that converts to LiOH and NH3 on reaction with H2O. It is majorly used in hydrogen storage applications.[1][2][3]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Analysis Note

Hydrogen content, XRD plots and metal purity data are available upon request.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mechanism for non-stoichiometry in the lithium amide/lithium imide hydrogen storage reaction
David WIF, et al.
Journal of the American Chemical Society, 129(6), 1594-1601 (2007)
Interaction between lithium amide and lithium hydride.
Chen P
The Journal of Physical Chemistry B, 107(39), 10967-10970 (2003)
First-principles study on lithium amide for hydrogen storage
Miwa K, et al.
Physical Review. B, Condensed Matter and Materials Physics, 71(19), 195109-195109 (2005)

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service