Skip to Content
Merck
All Photos(1)

Key Documents

684503

Sigma-Aldrich

[6,6]-Pentadeuterophenyl C61 butyric acid methyl ester

99.5%, 99.5 atom % D

Synonym(s):

1-[3-(Methoxycarbonyl)propyl]-1-pentadeuterophenyl-[6.6] C61, 3′-(Phenyl-d5) 3′H-cyclopropyl[1,9] [5,6]fullerene-C60-Ih-3′-butanoic acid methyl ester, d5-PCBM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72D5H9O2
CAS Number:
Molecular Weight:
915.91
UNSPSC Code:
12352103
NACRES:
NA.23

description

functionalized fullerene

isotopic purity

99.5 atom % D

Assay

99.5%

form

powder

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2(CCCC(=O)OC)[C]3=4c5c6c7c8c9c%10c(c%11c%12c3c%13c5c%14c%15c6c%16c7c%17c9c%18c%19c%10c%20c%11c%21c%12c%22c%13c%23c%14c%24c%15c%25c%16c%26c%17c%18c%27c%28c%19c%20c%29c%21c%30c%22c%23c%31c%24c%32c%25c%26c%27c%33c%28c%29c%30c%31c%32%33)[C]2=48

InChI

1S/C72H14O2/c1-74-11(73)8-5-9-70(10-6-3-2-4-7-10)71-66-58-50-40-30-22-14-12-13-16-20-18(14)26-34-28(20)38-32-24(16)25-17(13)21-19-15(12)23(22)31-37-27(19)35-29(21)39-33(25)43-42(32)52-46(38)56-48(34)54(44(50)36(26)30)62(66)64(56)68-60(52)61-53(43)47(39)57-49(35)55-45(37)51(41(31)40)59(58)67(71)63(55)65(57)69(61)72(68,70)71/h2-4,6-7H,5,8-9H2,1H3/i2D,3D,4D,6D,7D

InChI key

FIGVSQKKPIKBST-QRKCWBMQSA-N

General description

[6,6]-Pentadeuterophenyl C61 butyric acid methyl ester (PC60BM) is a deuterated fullerene that can be used as an acceptor molecule. It has a fullerene as the core and deuterium and benzyl as the attachment. The deuterium atoms facilitate an increase in the quantum efficiency of the electrochemical devices.

Application

PC60BM is a conjugating polymer that can be used as an alternative to the conventionally used PCBM for the organic electronic based applications, which include polymeric solar cells and organic light emitting diodes (OLEDs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of poly (p-phenylene vinylene)/methanofullerene blends of polymer solar cells by time-of-flight secondary ion mass spectrometry
Bulle-Lieuwma CWT, et al.
Applied Surface Science, 231(45), 274-277 (2004)
Bulle-Lieuwma, C.,; Van Duren, J. et al.
Applied Surface Science, 274, 231-232 (2004)
Manipulating the crystal structure of a conjugated polymer for efficient sequentially processed organic solar cells
Yi A, et al.
Nanoscale, 10(45), 21052-21061 (2018)
Deuterium isotope effect on bulk heterojunction solar cells. Enhancement of organic photovoltaic performances using monobenzyl substituted deuteriofullerene acceptors
Lu S, et al.
Organic Letters, 15(22), 5674-5677 (2013)

Articles

Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.

PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.

Find various photovoltaic and bioscience-based applications of fullerenes.

SWCNTs show promise in FETs, solar cells, and photodetectors due to their ultrafast charge transport mobility.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service