Skip to Content
Merck
All Photos(1)

Key Documents

683930

Sigma-Aldrich

2-Tributylstannylbenzo[b]thiophene

95%

Synonym(s):

Benzo[b]thien-2-yltributylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32SSn
CAS Number:
Molecular Weight:
423.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.5637

density

1.184 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1cc2ccccc2s1

InChI

1S/C8H5S.3C4H9.Sn/c1-2-4-8-7(3-1)5-6-9-8;3*1-3-4-2;/h1-5H;3*1,3-4H2,2H3;

InChI key

UJPVMOUQRNYNSA-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service