Skip to Content
Merck
All Photos(1)

Key Documents

682869

Sigma-Aldrich

(11bS,11′bS)-4,4′-(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin

Synonym(s):

S,S-Reetz X-Diphosphonite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H36O5P2
CAS Number:
Molecular Weight:
838.82
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]22/D -245°, c = 1 in chloroform

mp

195-207 °C (dec.)

SMILES string

CC1(C)c2cccc(P3Oc4ccc5ccccc5c4-c6c(O3)ccc7ccccc67)c2Oc8c(cccc18)P9Oc%10ccc%11ccccc%11c%10-c%12c(O9)ccc%13ccccc%12%13

InChI

1S/C55H36O5P2/c1-55(2)41-21-11-23-47(61-57-43-29-25-33-13-3-7-17-37(33)49(43)50-38-18-8-4-14-34(38)26-30-44(50)58-61)53(41)56-54-42(55)22-12-24-48(54)62-59-45-31-27-35-15-5-9-19-39(35)51(45)52-40-20-10-6-16-36(40)28-32-46(52)60-62/h3-32H,1-2H3

InChI key

DXZANFKSPAISOB-UHFFFAOYSA-N

Application

  • Catalyst for asymmetric hydrogenation of quinolines catalyzed by iridium complexes of BINOL-derived diphosphonites
  • Ligand in asymmetric Ni-catalyzed hydrocyanation of styrene and other vinylarenes
This diphosphonite ligand has been successfully employed in Noyori-type asymmetric transfer hydrogenation of prochiral ketones.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manfred T Reetz et al.
Journal of the American Chemical Society, 128(4), 1044-1045 (2006-01-26)
A BINOL-derived diphosphonite having a xanthene backbone is an excellent bidentate ligand in Ru-catalyzed 2-propanol-based transfer hydrogenation of aryl/alkyl and alkyl/alkyl ketones (ee = 76-99%). Even notoriously difficult ketones such as isopropyl methyl ketone are reduced with extraordinarily high enantioselectivity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service