Skip to Content
Merck
All Photos(2)

Documents

680516

Sigma-Aldrich

N-Boc-5-methoxy-2-indolylboronic acid

≥95%

Synonym(s):

N-(tert-Butoxycarbonyl)-5-methoxyindole-2-boronic acid, N-Boc-5-methoxyindole-2-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18BNO5
CAS Number:
Molecular Weight:
291.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

102-107 °C

storage temp.

2-8°C

SMILES string

COc1ccc2n(C(=O)OC(C)(C)C)c(cc2c1)B(O)O

InChI

1S/C14H18BNO5/c1-14(2,3)21-13(17)16-11-6-5-10(20-4)7-9(11)8-12(16)15(18)19/h5-8,18-19H,1-4H3

InChI key

PZLVPMBCKHDVKT-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant for:
  • Preparation of dihalogenated pyrazoles via dipolar cycloaddition by palladium-catalyzed cross-coupling processes
  • Suzuki-Miyaura cross-coupling
  • Synthesis of the isocryptolepine alkaloid and its related skeletons using a modified Pictet-Spengler reaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service