Skip to Content
Merck
All Photos(1)

Documents

680508

Sigma-Aldrich

N-(4-Bromophenyl)methanesulfonamide

97%

Synonym(s):

4-(Methylsulfonylamido)bromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NHSO2CH3
CAS Number:
Molecular Weight:
250.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

137-141 °C

SMILES string

CS(=O)(=O)Nc1ccc(Br)cc1

InChI

1S/C7H8BrNO2S/c1-12(10,11)9-7-4-2-6(8)3-5-7/h2-5,9H,1H3

InChI key

KKOIRAFXKFYZHQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sulfonyl chlorides are often chosen as building blocks in medicinal chemistry for their ability to easily react with heterocyclic amines to create complex sulfonamides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service