Skip to Content
Merck
All Photos(1)

Key Documents

678953

Sigma-Aldrich

4-(1-Piperidinyl)benzaldehyde

97%

Synonym(s):

1-(4-Formylphenyl)piperidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$747.00

R$747.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$747.00

About This Item

Empirical Formula (Hill Notation):
C12H15NO
CAS Number:
Molecular Weight:
189.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$747.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

61-64 °C

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(cc1)N2CCCCC2

InChI

1S/C12H15NO/c14-10-11-4-6-12(7-5-11)13-8-2-1-3-9-13/h4-7,10H,1-3,8-9H2

InChI key

ILJVPSVCFVQUAD-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Anti-inflammatory agents
  • Piperidine incorporated α-aminophosphonates for antibacterial agents
  • 5-Hydroxyaurone derivatives as growth inhibitors against HUVEC and some cancer cell lines
  • NR2B selective NMDA receptor antagonists
  • Fulleropyrrolidines
  • Fluorophores for monitoring polymerization processes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elitsa Pavlova et al.
European biophysics journal : EBJ, 44(8), 623-634 (2015-08-01)
The overproduction of free radicals and reactive oxygen species (ROS) has been proved as a basic damage mechanism and cause for oxidative stress. Their measurement is often hindered by the low signal. This could be resolved with the application of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service