Skip to Content
Merck
All Photos(1)

Documents

676101

Sigma-Aldrich

Ammonia-15N solution

98 atom % 15N, 7 M in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
15NH3
Molecular Weight:
18.02
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % 15N

concentration

7 M in methanol

mass shift

M+1

storage temp.

2-8°C

SMILES string

[15NH3]

InChI

1S/H3N/h1H3/i1+1

InChI key

QGZKDVFQNNGYKY-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service