Skip to Content
Merck
All Photos(2)

Key Documents

675245

Sigma-Aldrich

3-Pentylmagnesium bromide solution

2.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11BrMg
CAS Number:
Molecular Weight:
175.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

0.953 g/mL at 25 °C

SMILES string

CCC(CC)[Mg]Br

InChI

1S/C5H11.BrH.Mg/c1-3-5-4-2;;/h5H,3-4H2,1-2H3;1H;/q;;+1/p-1

InChI key

PLWIGERSUIOLRS-UHFFFAOYSA-M

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-20.2 °F

Flash Point(C)

-29 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service