Skip to Content
Merck
All Photos(2)

Documents

673978

Sigma-Aldrich

2,6-Bis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazolin-2-yl]pyridine

Synonym(s):

(3aS,3′aS,8aR,8′aR)-2,2′-(2,6-Pyridinediyl)bis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole, (3aS,8aR)-in-pybox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19N3O2
CAS Number:
Molecular Weight:
393.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]20/D -375°, c = 1 in dichloromethane (typical)

mp

276-281 °C

SMILES string

C1[C@H]2OC(=N[C@H]2c3ccccc13)c4cccc(n4)C5=N[C@@H]6[C@@H](Cc7ccccc67)O5

InChI

1S/C25H19N3O2/c1-3-8-16-14(6-1)12-20-22(16)27-24(29-20)18-10-5-11-19(26-18)25-28-23-17-9-4-2-7-15(17)13-21(23)30-25/h1-11,20-23H,12-13H2/t20-,21-,22+,23+/m1/s1

InChI key

BZSJUFJXCHHRHW-LUKWVAJMSA-N

Related Categories

Application

Reactant involved in synthesis of:
  • Chiral ruthenium(II)-pybox complexes for use as intramolecular C-H amination catalysts
  • Bis(oxazolinyl_pyridine-scandium(III) triflate complex catalyst for enantioselective addition of pyrroles to indoles
  • Chiral N,N,N-tridentate pybos and pyboxazine ligands

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service