Skip to Content
Merck
All Photos(1)

Documents

670650

Sigma-Aldrich

(1R,2R)-trans-N-Boc-1,2-cyclohexanediamine

97%

Synonym(s):

(1R)-trans-N-Boc-1,2-diaminocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O2
CAS Number:
Molecular Weight:
214.30
Beilstein:
7023564
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5%
97%

form

solid

optical activity

[α]/D -26±2°, c = 1 in chloroform

optical purity

ee: ≥99.0%

SMILES string

CC(C)(C)OC(=O)N[C@@H]1CCCC[C@H]1N

InChI

1S/C11H22N2O2/c1-11(2,3)15-10(14)13-9-7-5-4-6-8(9)12/h8-9H,4-7,12H2,1-3H3,(H,13,14)/t8-,9-/m1/s1

InChI key

AKVIZYGPJIWKOS-RKDXNWHRSA-N

Application

(1R,2R)-trans-N-Boc-1,2-cyclohexanediamine can be used as:
  • An organocatalyst in the intramolecular desymmetrization of cyclohexanones to yield 2-azabicyclo[3.3.1]nonane.
  • A starting material in the synthesis of a chiral nickel catalyst applicable for the Michael addition reaction of 1,3-dicarbonyl compounds to yield nitroalkenes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Desymmetrization of Prochiral Cyclohexanones by Organocatalytic Intramolecular Michael Additions to α,β-Unsaturated Esters
Gammack Y, et al.
Angewandte Chemie (International Edition in English), 54(16), 4899-4903 (2015)
Robust and Recyclable Self-Supported Chiral Nickel Catalyst for the Enantioselective Michael Addition
Bissessar D, et al.
advanced synthesis and catalysis, 358(12), 1982-1988 (2016)
Robust and Recyclable Self-Supported Chiral Nickel Catalyst for the Enantioselective Michael Addition
Bissessar D, et al.
Advanced Synthesis & Catalysis, 358(12), 1982-1988 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service