Skip to Content
Merck
All Photos(1)

Documents

670103

Sigma-Aldrich

2-(1,1-Dimethylpropyl)-6-(diphenylphosphino)pyridine

≥98.0% (HPLC)

Synonym(s):

2-tert-Amyl-6-(diphenylphosphino)pyridine, ALPYPHOS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24NP
CAS Number:
Molecular Weight:
333.41
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

78-82 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

CCC(C)(C)c1cccc(n1)P(c2ccccc2)c3ccccc3

InChI

1S/C22H24NP/c1-4-22(2,3)20-16-11-17-21(23-20)24(18-12-7-5-8-13-18)19-14-9-6-10-15-19/h5-17H,4H2,1-3H3

InChI key

ZEQMNGUCEYRQRF-UHFFFAOYSA-N

Application

Catalyst for:
  • Regioselective synthesis of oligo-1,4-diols
  • Anti-Markovnikov hydration

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurélie Labonne et al.
Organic letters, 8(25), 5853-5856 (2006-12-01)
The anti-Markovnikov hydration of terminal alkynes to give aldehydes is catalyzed by complexes derived in situ from air-stable [CpRu(eta6-naphthalene)]PF6 (C) and 6-aryl-2-diphenylphosphinopyridines (L). Ligands L are readily available from a modular synthesis. Increasing the size of the ligand C-6 aryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service